Hydrogenperoxid 34,5-36,5%, meets analytical specification of Ph. Nord.

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

18304H-5L 18304H-1L 18304H-6X1L
HONC18304H-5LEA 1340 DKK
HONC18304H-5L HONC18304H-1L HONC18304H-6X1L
Hydrogenperoxid 34,5-36,5%, meets analytical specification of Ph. Nord.
Hydrogen peroxide
Formel: H₂O₂
Molekylvægt: 34.01 g/mol
Kogepunkt: 150.2
Smeltepunkt: –33 °C
Massefylde: 1.110 g/cm3 (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011333
CAS-nummer: 7722-84-1
UN nummer: 2014
ADR: 5.1 II
Merck Index: 14,04798

Bestil nu

Specifikationer Testresultater

Assay 34.5 - 36.5 %
Non-volatile matter Max. 0.05 %
Residue on ignition Max. 0.03 %
Free acid (as H₂SO₄) Max. 0.02 %
Heavy metals (as Pb) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR