Rengøringsmidler, pulvere, rea-pur
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9710102 9710105
HIRS9710102EA 430 DKK
HIRS9710102 148-4233
Rengøringsmidler, pulvere, rea-pur
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Surface cleaner that rapidly cleans materials such as glass, porcelain, rubber, plastics and metals of oil, grease, wax, resin, silicone, blood, protein and serum residues. Also suitable for cleaning and decontamination of walls and floors. Environmentally friendly as the active agents are biologically degradable.
  • Graduations on glassware are unaffected
  • Free of chlorine and caustic alkalis
  • Good skin toleration and non toxic
  • Extremely economical: Typically 0,5 % working solution
  • Suitable for ultrasonic cleaning baths
Advarsel: Long exposure timesMore Product Information