Burettes, Pellet pattern, brown, lateral stopcock
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
342016060 342017060 342017560 342036060 342037060 342037560
HIRS342016060EA 1910 DKK
HIRS342016060 HIRS342017060 HIRS342017560 HIRS342036060 HIRS342037060 HIRS342037560
Burettes, Pellet pattern, brown, lateral stopcock
Buretter
Automatic burettes, Pellet pattern, class B, DURAN® brown glass, with white graduation.
  • Schellbach stripes for enhanced legibility
  • Lateral stopcock with glass key or PTFE spindle
Certificeringer: Compliance with DIN EN ISO 385.