Avantor® Alltima HP, HPLC Columns, C18-HL, 5 µm
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HICH87699EA 8530 DKK
HICH87699 HICH87700 HICH87697 HICH87698 HICH5135351 87624/N 87701/N
Avantor® Alltima HP, HPLC Columns, C18-HL, 5 µm
Kromatografikolonner
Avantor® Alltima HP columns contain the highest carbon load for superior retention and loadability.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
  • Low to no detectable column bleed
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.