Avantor® Hichrom HI-5, GC Columns
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HI02-05-005-2EA 3320 DKK
HI02-05-005-2 HI02-05-005-2.5 HI02-10-020-10 HI02-10-020-15 HI02-10-020-5 HI02-10-040-10 HI02-10-040-15 HI02-15-015-10 HI02-25-025-15G5 HI02-25-025-30G5 HI02-25-025-60G5 HI02-25-100-30G5 HI02-32-100-30G5 HI02-53-050-30G10 HI02-53-100-30G5 HI02-18-018-20 HI02-18-018-40 HI02-18-018-50 HI02-18-020-10 HI02-18-020-40 HI02-18-020-50 HI02-18-020-60 HI02-18-040-40 HI02-20-010-20 HI02-20-020-50 HI02-20-020-60 HI02-20-040-10 HI02-20-040-20 HI02-20-040-40 HI02-20-040-60 HI02-20-100-10 HI02-20-100-20 HI02-20-100-40 HI02-20-100-50 HI02-20-100-60 HI02-25-010-30 HI02-25-010-50 HI02-25-025-105 HI02-25-025-30 HI02-25-025-60 HI02-25-050-105 HI02-25-050-50 HI02-25-100-10 HI02-25-100-105 HI02-25-100-15 HI02-25-100-50 HI02-32-010-105 HI02-32-010-15 HI02-32-010-60 HI02-32-025-105 HI02-32-025-15 HI02-32-025-30 HI02-32-025-60 HI02-32-050-10 HI02-32-050-105 HI02-32-050-60 HI02-32-100-105 HI02-32-100-50 HI02-32-150-105 HI02-32-150-60 HI02-32-300-10 HI02-32-300-105 HI02-32-300-50 HI02-32-300-60 HI02-32-500-105 HI02-32-500-30 HI02-32-500-50 HI02-53-010-10 HI02-53-010-15 HI02-53-010-30 HI02-53-010-60 HI02-53-025-15 HI02-53-025-30 HI02-53-025-60 HI02-53-050-15 HI02-53-050-30 HI02-53-050-50 HI02-53-050-60 HI02-53-100-10 HI02-53-100-15 HI02-53-100-50 HI02-53-150-10 HI02-53-150-30 HI02-53-150-60 HI02-53-150-75 HI02-53-300-50 HI02-53-300-75 HI02-53-500-10 HI02-53-500-30 HI02-53-500-50 HI02-32-500-25 HIGCHI0232025155IN HIGCHI02-32-050-25 HIGCHI02-32-017-25 HIGCHI02-32-012-50 HIGCHI02-32-052-50
Avantor® Hichrom HI-5, GC Columns
Kromatografikolonner
Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 is a crossbonded phase and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded 5% phenyl - 95%Methyl Polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.