Ultra low temperature freezers, with 5 inner doors, –86 °C, TDE series
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TDE30086FV5I-U TDE40086FV5I-U TDE50086FV5I-U TDE60086FV5I-U
HERATDE30086FV5I-UEA 98850 DKK
HERATDE30086FV5I-U HERATDE40086FV5I-U HERATDE50086FV5I-U HERATDE60086FV5I-U
Ultra low temperature freezers, with 5 inner doors, –86 °C, TDE series
Frysere Ultra lav temperatur frysere
Thermo Scientific™ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models with 5 Inner doors, maximizing storage capacity from 30000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab. Sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-