Racks for sliding drawer freezer, Matrix, Matrix 2D and Nunc CryoBank
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1950815 1950799 1950789 1950819 1950787
HERA1950815EA 1570 DKK
HERA1950815 HERA1950799 HERA1950789 HERA1950819 HERA1950787
Racks for sliding drawer freezer, Matrix, Matrix 2D and Nunc CryoBank
Stativer Stativer til Kryobokse
Maximise tube storage capacity per freezer with Thermo Scientific™ Sliding drawer racks for tubes (4 inner door freezers), a racking solution for four inner-door, ultra-low temperature freezers. Use in conjunction with Thermo Scientific™ dense storage vials.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country
  • Material: Stainless steel
  • Colour: Silver
Thermo Scientific freezer racking and tubing solutions offer a convenient, streamlined process to maximise sample storage in yourMore Product Information