PharmaFocus® Premium Silicone Tubing
Leverandør: Freudenberg Medical Europe GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
49464653 49447156 49464654 49463297 49450058 49464655 49464656 49464659 49450926 49447157 49464686 49449004 49457002 49464687 49464689 49447181 49464690 49464692 49462201 49443817 49464531 49464693 49464694 49443813 49464700 49464701 49464702 49464706 49464707 49464708 49464703 49464704 49464705 49449005
HELI49464653EA 1320 DKK
HELI49464653 HELI49447156 HELI49464654 HELI49463297 HELI49450058 HELI49464655 HELI49464656 HELI49464659 HELI49450926 HELI49447157 HELI49464686 HELI49449004 HELI49457002 HELI49464687 HELI49464689 HELI49447181 HELI49464690 HELI49464692 HELI49462201 HELI49443817 HELI49464531 HELI49464693 HELI49464694 HELI49443813 HELI49464700 HELI49464701 HELI49464702 HELI49464706 HELI49464707 HELI49464708 HELI49464703 HELI49464704 HELI49464705 HELI49449005
PharmaFocus® Premium Silicone Tubing
Slanger Overførselsslange
Designed to meet a wide range of performance requirements in pharmaceutical applications. PharmaFocus® Premium Silicone Tubing is ideal for demanding fluid transfer processes and laboratory use. These products have been specifically formulated for use with sensitive media found in pharmaceutical and biotech processing.
  • Performs in high purity fluid transfer and pump applications
  • Tube resilience and adaptability due to excellent flexibility
  • High visibility of fluid transport
  • Versatile processing with cut-to-length, spools and packaging
  • Biocompatibility for