Hei-PLATE Mix 'n' Heat Core+ Magnetic Hotplate Stirrers
Leverandør: Heidolph Instruments GmbH & Co.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
506-22100-00 506-21100-00 506-21150-00 506-21151-00 506-21155-00 506-21156-00 506-21151-03 506-22150-00 506-22151-00
HEID506-22100-00EA 8050 DKK
HEID506-22100-00 HEID506-21100-00 HEID506-21150-00 HEID506-21151-00 HEID506-21155-00 HEID506-21156-00 HEID506-21151-03 HEID506-22150-00 HEID506-22151-00
Hei-PLATE Mix 'n' Heat Core+ Magnetic Hotplate Stirrers
Omrørere Magnetomrørere med varme
Magnetic stirrers with heating function for academic standards with temperature sensor.
  • The controller’s locking function protects against accidental adjustment
  • The values in the illuminated display are easy to read even from a distance
  • Activated functions are easily detectable due to the LED rings
  • Continuous mode of operation
  • Connection L+N+PE
  • LCD display
Precise setting options and monitoring of the device parameters via the digital display and connection for the external Pt 1000 temperature sensor. Precise values without temperature fluctuationsMore Product Information