Laboratory tray sets
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42002008
HECH42002008EA 1410 DKK
HECH42002008
Laboratory tray sets
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Complete laboratory tray set consists of plastic tray, Plexiglas® stand, 2 wide neck bottles (30 ml) and 1 narrow neck bottle (50 ml).