Nissel tubes
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41145010
HECH41145010EA 188 DKK
HECH41145010
Nissel tubes
Kliniske diagnose systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Glass.
  • Fused blue graduation
For the volumetric determination of fibrin. With directions for use - "heat fibrin according to Schulz".