Counting chambers, Jessen
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40450702
HECH40450702EA 801 DKK
HECH40450702
Counting chambers, Jessen
Objektglas Tællekamre
These double ruling chambers are used for blood tests, urine analysis, and counting cells in cerebrospinal fluid (CSF).