Pipette workstation
Leverandør: HEATHROW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HS234631
HEATHS234631EA 1820 DKK
HEATHS234631
Pipette workstation
Pipettestativer
Keep pipette and disposable tips together in one convenient stand.
  • Holds up to four pipettes on top shelf
  • Dual bins hold multiple size tips
  • Self-closing lid provides dust-free storage
  • Oversized lip allows for easy access when wearing gloves
Acrylic workstation is opaque on sides with clear front and bin lids for easy content identification.