Pipette controller, manual, RF1000
Leverandør: HEATHROW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HS1000RFU
HEATHS1000RFUEA 769 DKK
HEATHS1000RFU
Pipette controller, manual, RF1000
Pipetter Pipettehjælpere
The RF1000 pipette controller is lightweight and easy to use. It is a manual pipette controller and is uniquely designed for one-handed operation.
  • Tapered silicone rubber pipette adapter allows a large range of pipette sizes ranging 1 to 100 ml (replaceable when worn)
  • Filter: PTFE/25 mm Ø / 0,45 μm filter in PP housing
  • Rubber pipette adapter replaceable when worn
  • Stay organized with two additional nose cones (green, white) for colour identification
  • Includes one replaceable 0,45 µm hydrophobic filter to protect your samples and internal components