Lids for 96-well PCR plates, ComfortLid
Leverandør: HAMILTON ROBOTICS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
814300
HAMR814300EA 2941.42 DKK
HAMR814300
Lids for 96-well PCR plates, ComfortLid
Låg Mikroplade låg
Gentle pressure is applied to the ComfortLid to seal PCR plates and prevent evaporation and contamination, used in equipped liquid handling workstations.
  • Free of DNase, RNase, pyrogen (endotoxins), human DNA, PCR inhibitors
  • Made of polycarbonate (PC) and thermoplastic elastomers (TPE)
These cost-effective and disposable lid has a stackable design that can be transported by all robotic transport arms.
Emballering: Packed as 10 lids/blister and 5 blister/box.