Syringe rack
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
204880
HAMI204880EA 2680 DKK
HAMI204880
Syringe rack
Stativer Syringe Racks
This brushed stainless steel rack is used for clean and safe storage of syringes with plastic holder. A lip on the plastic holder protects the syringe against accidental touching and falling down. The distance of the plastic holder prevents the syringe from getting in contact with the metal plate.
  • A trough underneath prevents the soiling of worplace with liquid
  • Usable as free standing or wall model