Electrode cleaning solution, RENOVO.X
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S16M002
HACHS16M002EA 216 DKK
HACHS16M002
Electrode cleaning solution, RENOVO.X
Elektrolytter Opløsninger til Elektrode opbevaring
This extra strong electrode cleaning solution is ideal for use when stronger cleaning agents are required, for example when the solutions contain protein.