Conductivity sensor, inductive, 3700 SC
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D3708E2T.99
HACHD3708E2T.99EA 0 DKK
HACHD3708E2T.99
Conductivity sensor, inductive, 3700 SC
Elektroder Elektroder til ledningsevnemåling
The innovative technology of Hach’s inductive conductivity sensors eliminates polarization and electrode coating problems in harsh environments. The sensor comes complete with standard sensor cable length of 6 m, digital gateway, and a 1 m digital extension cable.
  • Sanitary (CIP) flange style: 2-inch flange, special cap, and EPDM compound gasket
  • Conforms to provisions of 3-A Sanitary Standards