Kaliumchlorid ACS

Leverandør: Hach
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassiumchloride

76401
HACH76401EA 324 DKK
HACH76401
Kaliumchlorid ACS
Potassium chloride
Formel: KCl
Molekylvægt: 74.55 g/mol
Kogepunkt: 1420
Smeltepunkt: 773
MDL Number: MFCD00011360
CAS-nummer: 7447-40-7
Merck Index: 14,07621

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR