Kaliumiodid solution_SC 30 g/L

Leverandør: Hach
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Dipotassium diiodide, Kali iodide

34332
HACH34332EA 203 DKK
HACH34332
Kaliumiodid solution_SC 30 g/L
Potassium iodide

Reagent for use in iodometric titrations.


Method: 8210

Formel: Kl
Molekylvægt: 166 g/mol
Kogepunkt: 1330
Smeltepunkt: 686
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011405
CAS-nummer: 7681-11-0
Merck Index: 13,07727

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR