4,5-Difluor-2-nitroanilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4-Difluoro-6-nitroaniline

H55830.03 H55830.06
H55830.03EA 690 DKK
H55830.03 H55830.06
4,5-Difluor-2-nitroanilin ≥98%
4,5-Difluor-2-nitroanilin

Purity: 98% CAS No.: 78056-39-0 Molecular Formula: C6H4F2N2O2 Formula Weight: 174.11 Melting Point: 107-108° Beilstein Reference: 2723242

Formel: C₆H₄F₂N₂O₂
Smeltepunkt: 107…108 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010876
CAS-nummer: 78056-39-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR