2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H54088.MD H54088.03 H54088.06
H54088.MDEA 811 DKK
H54088.MD H54088.03 H54088.06
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Purity: 97% CAS No.: 306960-77-0 Molecular Formula: C6H3F3N2O2 Formula Weight: 192.10 Melting Point: 170-175° MDL No.: MFCD09054851

Formel: C₆H₃F₃N₂O₂
Smeltepunkt: 170…175 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09054851
CAS-nummer: 306960-77-0

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR