Methyl (4-fluorophenyl)acetate ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 2-(4-fluorophenyl)acetate

H32236.06 H32236.14
H32236.06EA 553 DKK
H32236.06 H32236.14
Methyl (4-fluorophenyl)acetate ≥98%
Methyl (4-fluorophenyl)acetate

Purity: 98% CAS No.: 34837-84-8 Molecular Formula: C9H9FO2 Formula Weight: 168.17 MDL No.: MFCD00800608

Formel: C₉H₉FO₂
Molekylvægt: 168.17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00800608
CAS-nummer: 34837-84-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR