Diethyl-4,4-difluoroheptanedioate 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Diethyl 4,4-difluoroheptanedioate

H31544.03
H31544.03EA 1590 DKK
H31544.03
Diethyl-4,4-difluoroheptanedioate 97%
Diethyl-4,4-difluoroheptanedioate

Purity: 97% CAS No.: 22515-16-8 Molecular Formula: C11H18F2O4 Formula Weight: 252.26 Refractive Index: 1.4150 Boiling Point: 108°/3mm MDL No.: MFCD08146634

Formel: C₁₁H₁₈F₂O₄
Kogepunkt: 108 °C (3 mmHg)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08146634
CAS-nummer: 22515-16-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR