2,3-Difluor-6-methoxybenzoesyre ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H26830.03 H26830.09
H26830.03EA 824 DKK
H26830.03 H26830.09
2,3-Difluor-6-methoxybenzoesyre ≥97%
2,3-Difluor-6-methoxybenzoesyre

Purity: 97% CAS No.: 773873-26-0 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 126-129° MDL No.: MFCD04116018

Formel: C₈H₆F₂O₃
Smeltepunkt: 126…129 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04116018
CAS-nummer: 773873-26-0
EINECS: None

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR