Waterproof pH electrode, GE 114-WD
Leverandør: Greisinger
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
610460
GRSG610460EA 0 DKK
GRSG610460
Waterproof pH electrode, GE 114-WD
Elektroder pH-elektroder
The GE114 is a waterproof and universally applicable, robust and low-maintenance gel-electrode with pellon-diaphragm. It can be submersed for a longer time beyond the shaft.
  • Fully submersible
  • Pressure resistant
  • Low-maintenance
  • Universal and fast
  • GMH 35xx, GMH 55xx, G 7500 and G150x
  • Also for transmitter GPHU 014
Completely waterproof due to the special BNC-plug in combined with GMH55xx, G 7500 or G150x for demanding field application. Especially suited for fish farming, aquaristics (including salt water), drinking water, swimming pool and much more.