Cell culture dishes, square, CELLSTAR®
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
688160_240
GREI688160_240EA 3110 DKK
GREI688160_240
Cell culture dishes, square, CELLSTAR®
Dyrkningsplader Celledyrkningsskåle
PS, cell culture-treated, sterile.
  • Physical surface treatment for improved cell adhesion
  • Vents ensure efficient gas exchange
  • Certified free from RNase, DNase and human DN
  • Non pyrogenic and non cytotoxic