Anti-Protein C -tag Antibody (HPC4) Rabbit Polyclonal Antibody

Leverandør: Genscript
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A00637-40
GENSA00637-40EA 1300 DKK
GENSA00637-40
Anti-Protein C -tag Antibody (HPC4) Rabbit Polyclonal Antibody
Antistoffer
Type: Tag
Antigen: Protein C -tag Antibody (HPC4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestil nu

Specifikationer

Environmentally Preferable
Antigen name Protein C-tag
Antigen synonymer Vitamin K-dependent protein C|PROC|Heavy chain of human protein C|Vitamin K-dependent protein C heavy chain|Vitamin K-dependent protein C light chain|Autoprothrombin IIA|Anticoagulant protein C|EDQVDPRLIDGK|Blood coagulation factor XIV|Cleaved into the following 3 chains
Antistof type Tag
Clonality Polyclonal
Konjugation Unconjugated
Reactivity
Vært Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Opbevaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR