Empty FPLC columns, G-TRAP™

Leverandør: G-Biosciences
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

786-1291 786-1290
GENO786-1291EA 460 DKK
GENO786-1291 GENO786-1290
Empty FPLC columns, G-TRAP™
Kromatografikolonner
These empty columns are designed for fast protein liquid chromatography.

  • Frit material is ultra-high molecular weight polyethylene

Columns are made of polypropylene and come in 1 ml and 5 ml sizes.

Leveringsinformation: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).
Bestil nu

Specifikationer

Environmentally Preferable
Maks. tryk 3 bar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR