Avantor® Genesis, HPLC Columns, C8
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FK5962EEA 6460 DKK
FK5962E FM5964 FM10962E FM15962E FM25962E FH1962-2/N GEN-5C8-250D FM25964E GEN-5C8-150D FK25964E HICHGEN-4C8-125DF
Avantor® Genesis, HPLC Columns, C8
Kromatografikolonner
Avantor® Genesis phases are based on high purity metal-free spherical silica. They are suitable for the analysis of a wide range of analytes.
  • Excellent peak shape and efficiency
  • Long column life