Hand disinfectants, Swarfega® Orange
Leverandør: Deb-STOKO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SOR2LT SORC4LTR
EVONSOR2LTEA 707 DKK
EVONSOR2LT EVONSORC4LTR
Hand disinfectants, Swarfega® Orange
Hudpleje Hånddesinfektion Liquid and Foam Skin Disinfectants
This is a solvent-free workshop cream with plastic microbeads, effective against strong and adherent soiling such as greases, oils and fuels.
  • Glycerin and wheat germ oil increase skin tolerance and hydrate the skin
  • Fresh fragrance based on natural orange extracts
  • Solvent-free form helps to prevent skin dryness
  • Contains glycerin and wheat germ extracts to help hydration of cutaneous skin
Contains micropolymer granules which provide deep cleaning and gently loosen crusted contaminants without damaging the skin. This rich and creamy lotion rinses offMore Product Information