Anti-Choline acetyltransferase Goat Polyclonal Antibody

Leverandør: Everest Biotech
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EB06636
EVBIEB06636EA 2530 DKK
EVBIEB06636
Anti-Choline acetyltransferase Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Choline acetyltransferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR