Centrifuges, bench top, refrigerated, 5920 R (General Lab Product)
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5948000.010 5948000.210 5948000.315 5948000.410 5948000.510 5948000.810 5948000.911 5948000.174
EPPE5948000.010EA 96200 DKK
EPPE5948000.010 521-2613 521-2614 521-2629 EPPE5948000.510 EPPE5948000.810 521-2774 521-2791
Centrifuges, bench top, refrigerated, 5920 R (General Lab Product)
Centrifuger Bordcentrifuger
This centrifuge features a superior capacity of up to 4×1000 ml which makes it the ideal instrument for high-throughput and large volume applications. It has a powerful refrigeration system with advanced temperature management to keep samples safe.
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • FastTemp function for quick pre-cooling; FastTemp pro® function allows for automatic precooling based on pre-programmable time and date
  • Continuous cooling maintains a constant temperature after the run has been completed – samples