Sterile Serological Pipettes
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030127.692 0030127.706 0030127.714 0030127.722 0030127.730 0030127.749
EPPE0030127.692EA 1220 DKK
EPPE0030127.692 EPPE0030127.706 EPPE0030127.714 EPPE0030127.722 EPPE0030127.730 EPPE0030127.749
Sterile Serological Pipettes
Pipetter Serologiske pipetter
Eppendorf Serological Pipettes ensure top-tier sterility and safety for sensitive lab applications. They are sterile, free from pyrogens, DNase, RNase, human and bacterial DNA, and non-cytotoxic. Ideal for cell culture and molecular biology tasks.
  • Clear and precise graduations for easy volume determination
  • Color-coded for easy identification of pipet sizes
  • Subunits with dispenser option to keep stored products safe
  • Individually wrapped for reliable sterility
  • Ultra-pure virgin polystyrene meets the requirement of USP VI
  • Harmonized and well-fitting operation