ACTH ELISA kit, grow cells
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ENZKIT1380001
ENZOENZKIT1380001EA 5450 DKK
ENZOENZKIT1380001
ACTH ELISA kit, grow cells
Assays ELISA'er
Highly sensitive ACTH ELISA kit enabling detection of ACTH in human plasma in 4.5 hours.
  • Sensitive measurement of ACTH, detecting as little as 0,46 pg/ml.
  • Negligible reactivity to similar hormones.
  • High throughput with results in 4.5 hours.
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ACTH in plasma in human samples.
The ACTH ELISA kit is a colorimetric immunoassay kit with results in 4,5 hours.