Anti-CD39 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AC2.5]

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ENZABS2360100
ENZOENZABS2360100EA 5110 DKK
ENZOENZABS2360100
Anti-CD39 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AC2.5]
Antistoffer
Recommended Applications: Flow Cytometry

Species reactivity: Human

Type: Primary
Antigen: CD39
Clonality: Monoclonal
Clone: AC2.5
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR