Cell extract HL60 (non-induced)
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BMLSW1010200
ENZOBMLSW1010200EA 386 DKK
ENZOBMLSW1010200
Cell extract HL60 (non-induced)
Elektroforese reagenser Western Blotting-reagenser
Whole-cell extract of human HL-60 leukemia cells.
  • Band with MW=116 kDa, which represents intact PARP, is detected by Western blotting
  • Ready to use for SDS-PAGE
  • Use approximately 20 µl sample per lane (~75,000 cells)