Hinokitiol ≥98% (by GC)

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: beta-Thujaplicine, 2-Hydroxy-4-isopropylcyclohepta-2,4,6-trien-1-one

BMLFR1100250
ENZOBMLFR1100250EA 2010 DKK
ENZOBMLFR1100250
Hinokitiol ≥98% (by GC)
Hinokitiol

Chelator that can induce differentiation in tumor cells.


Hinokitiol (β-thujaplicin) is a potent metal chelator that induces differentiation and apoptosis in teratocarcinoma F9 cells and tyrosinase, most likely via metal chelation.

Formel: C₁₀H₁₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00059582
CAS-nummer: 499-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15762b3734d109d4cce2622d1e9f477c1c80559c&vid=3c5c8a9e754392c15bce0bc3f3b196a8534ad6d0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity ≥98% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR