12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

BMLD1220050
ENZOBMLD1220050EA 2420 DKK
ENZOBMLD1220050
12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE)

Arachidonic acid metabolite.


Transcellular biosynthesis via interaction between platelets and neutrophils.

Formel: C₂₀H₃₂O₄
Opbevaringstemperatur: Ultra-lav temperatur fryser (–80°C)
CAS-nummer: 89614-44-8
EINECS: 200-578-6
UN nummer: 1170
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19a78c6bd5bc2b296cda612ce8f7e33c27d3c5f3&vid=21d2a02100e456759c41bc6a8f8c50493749e261&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Concentration 50 µg/ml
Formulation Oil dissolved in ethanol
Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR