Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BMLAK4120001
ENZOBMLAK4120001EA 4720 DKK
ENZOBMLAK4120001
Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
Assays Enzym Assays
A QuantiZyme™ Assay System. The MMP-13 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-13 (MMP-13, collagenase-3) using a thiopeptide as a chromogenic substrate.
Leveringsinformation: Kit contains MMP-13 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.