SDMA (human) ELISA kit
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALX850331KI01
ENZOALX850331KI01EA 13130 DKK
ENZOALX850331KI01
SDMA (human) ELISA kit
Assays ELISA'er
For the quantitative determination of symmetric dimethylarginine (SDMA) in human EDTA-plasma and serum. Does not cross-react with ADMA, NMMA or L-arginine.