Anti-Poly(Adp-Ribose) Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ALX804220R100
ENZOALX804220R100EA 4380 DKK
ENZOALX804220R100
Anti-Poly(Adp-Ribose) Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG3

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes poly(ADP-ribose) synthesized by a broad range of PARPs (poly(ADP-ribose) polymerases) like human, mouse, rat or Drosophila PARP enzyme.

Type: Primary
Antigen: POLY(ADP-RIBOSE)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
New Product
Bestil nu

Specifikationer

Antigen symbol PARP
Environmentally Preferable
Antigen name Poly(Adp-Ribose)
Antistof type Primary
Clonality Monoclonal
Reactivity
Vært Mouse
Isotype IgG3
Western blot Yes
ImmunoChemistry Yes
Flowcytometri Yes
Form Liquid. In 50 mM HEPES, pH 7.4, containing 100 mM sodium chloride, 1% BSA and 0,02% sodium azide.
Purification ≥95% (SDS-PAGE)
Koncentration 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR