Bovine native Ganglioside GM1 . sodium salt (from Brain)
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALX302001M001 ALX302001M005 ALX302001M010
ENZOALX302001M001EA 754 DKK
ENZOALX302001M001 ENZOALX302001M005 ENZOALX302001M010
Bovine native Ganglioside GM1 . sodium salt (from Brain)
Proteiner
Specific receptor for cholera toxin.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0b2c22505e4eb80961832ef31b0911a19d06582&vid=5c953c0b68328789696c739015d07d2cdff62037&prid=web&clid=SAPDAM