Anti-Glucocorticoid Receptor Alpha Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ALX210277C050
ENZOALX210277C050EA 6780 DKK
ENZOALX210277C050
Anti-Glucocorticoid Receptor Alpha Rabbit Polyclonal Antibody
Antistoffer
Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation, Western Blot, ChIP

Recognizes human, mouse and rat GRalpha. Detects a band of ~94kDa by Western blot.

Reconstitute with PBS.

Type: Primary
Antigen: GLUCOCORTICOID RECEPTOR ALPHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR