NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC)

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: H-Arg(di-Me)-OH.2HCl (asymmetrical), ADMA.2HCl

ALX106006M025
ENZOALX106006M025EA 1240 DKK
ENZOALX106006M025
NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC)
NG, NG-Dimethyl-L-arginine dihydrochloride

NOS inhibitor.


Endogenous nitric oxide synthase (NOS) inhibitor.


Hygroscopic and soluble in water (50 mg/ml).


Advarsel: Research use only.

Formel: C₈H₁₈N₄O₂·2HCl·H₂O
Opbevaringstemperatur: Fryser
CAS-nummer: 220805-22-1
UN nummer: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c4a77b1a87eb031727902beaa0ea8a744132396&vid=a8e6cd5b2aa0b40b8db5d01a8302fb1a28d94183&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity ≥97% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR