Protein A ELISA Kit
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ADI900057
ENZOADI900057EA 5320 DKK
ENZOADI900057
Protein A ELISA Kit
Assays ELISA'er
Most sensitive (9,01pg/ml) colorimetric ELISA for monitoring residual contamination.
For the quantitative determination of residual protein A contamination in protein A purified IgG preparations from any species.
The Protein A EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ultra-sensitive quantification (< 1 ppm) of Protein A residuals in purified humanized monoclonal antibody preparations. This kit recognizes four different Protein A constructs.
Bestillingsoplysninger: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate,Standard, TMB Substrate, Stop solution 2.