Pipette tracker, LightOne™ Illuminator Pro, Embitec®
Leverandør: EMBITEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LI-2100
EMBILI-2100EA 12350 DKK
EMBILI-2100
Pipette tracker, LightOne™ Illuminator Pro, Embitec®
Lysbord LED Transilluminatorer
Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 384-well or 96-well plates.
  • Red LEDs minimize quenching of fluorescent dyes, and light up in three distinct intensities
  • Auto and manual/hands-free advance modes
  • Dedicated LED for each sample well to provide accurate guidance
  • Auto and manual advance, or import plate design via USB connector
  • Online plate design generator available for creating sample layout file for importing
  • Material: White polycarbonate
The LightOne™ IlluminatorMore Product Information