o-Cresol

Leverandør: EHRENSTORFER
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: o-Methylphenol

CA15140200
EHERCA15140200EA 649 DKK
EHERCA15140200
o-Cresol
o-Cresol
Formel: C₇H₈O
Kogepunkt: 31 °C
Flammepunkt: 81 °C
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00002226
CAS-nummer: 95-48-7
UN nummer: 2076
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR