4-tert-Butyl-2-Chlorphenol

Leverandør: EHRENSTORFER
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(tert-Butyl)-2-chlorophenol

C10931125
EHERC10931125EA 1660 DKK
EHERC10931125
4-tert-Butyl-2-Chlorphenol
4-tert-Butyl-2-Chlorphenol
Formel: C₁₀H₁₃ClO
Molekylvægt: 184.66 g/mol
MDL Number: MFCD00019992
CAS-nummer: 98-28-2
EINECS: 202-652-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR