2-Amino-5-methylbenzenesulphonic acid

Leverandør: EHRENSTORFER
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Aminotoluene-3-sulphonic acid

C10228000
EHERC10228000EA 649 DKK
EHERC10228000
2-Amino-5-methylbenzenesulphonic acid
2-Amino-5-methylbenzenesulphonic acid
Formel: C₇H₉NO₃S
MDL Number: MFCD00007908
CAS-nummer: 88-44-8
EINECS: 201-831-3
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR